missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acetonitrile (ACN), LC-MS Grade

Product Code. 13474279 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
4 x 1 L
Unit Size:
1 L
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13474279 4 x 1 L Pack of 4
11581094 1 L 1 L
10501014 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13474279 Supplier Thermo Scientific™ Supplier No. 85188

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

An LC/MS-grade preparation with high purity and low UV-absorptivity that makes it suitable for HPLC and mass spectrometry applications.

Thermo Scientific™ Acetonitrile (ACN) is an LC/MS-grade preparation with high purity and low UV-absorptivity that makes it suitable for HPLC and mass spectrometry applications.

Thermo Scientific LC/MS Grade Acetonitrile is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance to provide you with the most sensitive detection across all wavelengths. Acetonitrile is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen atmosphere with PTFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • 37 quality tests ensure low, stable baselines and lot-to-lot consistency
  • Low UV absorbance yields low baselines and high detection sensitivity
  • Low impurities protects columns and simplifies analysis by eliminating extraneous peaks
  • Specially purified and tested to the highest specifications to ensure data integrity, maximize assay sensitivity, and to prolong equipment life
  • Store between 2°C to 25°C in a dry, cool, well-ventilated area, away from incompatible materials

Product Details:

  • Alternative names: Methyl cyanide, cyanomethane, ethanenitrile
  • Molecular formula: CH3CN
  • Molecular weight: 41.05
  • Density: 0.780 g/mL
TRUSTED_SUSTAINABILITY

Specifications

Grade LC-MS
Item Name Acetonitrile (ACN)
Quantity 4 x 1 L
Content And Storage Store between 2 to 25°C in a dry, cool, well-ventilated area, away from incompatible materials.
Form Liquid
Reagent Type Solvent (Pure)
Product Line Pierce
For Use With HPLC, Mass Spectrometry
CAS 75-05-8
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile (ACN), LC-MS Grade
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: acetonitrile

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.