missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Top 2 Abundant Protein Depletion Spin Columns

Product Code. 13414319 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
24 Columns
6 Columns
Unit Size:
Pack of 24
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13414319 24 Columns Pack of 24
13404319 6 Columns Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13414319 Supplier Thermo Scientific™ Supplier No. 85162

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Decrease the abundant albumin and antibody components of human plasma samples in preparation for mass spectrometry or 1D and 2D gel electrophoresis.

Decrease the abundant albumin and antibody components of human plasma samples in preparation for mass spectrometry or 1D and 2D gel electrophoresis with Thermo Scientific™ Pierce Abundant Protein Depletion Spin Columns.

Additional Information and Online Ordering

The Pierce Top 2 Abundant Protein Depletion Spin Columns use highly specific immobilized anti-HSA and anti-IgG antibodies to remove human serum albumin (HSA) and all major subclasses of gamma globulin (IgG) from serum, plasma or spinal fluids. Similarly, the Pierce Top 12 Abundant Protein Depletion Spin Columns are designed to remove HSA, IgG and 10 other high-abundance proteins from human serum or plasma. The resins are provided in an economical and convenient spin column specifically designed one step processing and for single use.

Highlights:

  • Optimized – resin in spin columns is scaled and optimized for treating 10μL (600μg) of human plasma samples for mass spectrometry and/or 1D and 2D electrophoresis

  • Efficient – kits remove >90% of IgG and >95% of albumin, plus other abundant proteins (Top 12)

  • Fast – process samples in 40 to 60 minutes (depending on resin)

  • Economical – cost effective spin columns are priced for single use

  • Reduce variability – One time use prevents abundant protein carryover and experimental variability

  • Two resin options – choose the system appropriate for your need: 2 or 12 protein depletion columns

Top 2 Spin Columns Remove:

  • Albumin
  • IgG

Top 12 Spin Columns Remove:

  • Albumin
  • IgG
  • α1-Acid Glycoprotein
  • α1-Antitrypsin
  • α2-Macroglobulin
  • Apolipoprotein A-I
  • Apolipoprotein A-II
  • Fibrinogen
  • Haptoglobin
  • IgA
  • IgM
  • Transferrin
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Microcentrifuge, Mass Spectrometer
Product Type Spin Column
Product Line Pierce
Content And Storage Store at 4°C—do not freeze
Starting Material Plasma, Serum
Final Product Type Protein
Detection Method Mass Spectrometry
Workflow Step Sample Enrichment, Sample Clean Up
Quantity 24 Columns
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.