missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ 6xHis Protein Tag Stain Reagent Set

Product Code. 10322234 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10322234 1 L 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10322234 Supplier Thermo Scientific™ Supplier No. 24570

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Preferentially detect histidine-tagged (His-tagged) recombinant fusion proteins in polyacrylamide gels with this fluorescent stain.

Thermo Scientific™ Pierce 6xHis Protein Tag Stain Kit is a fluorescent stain for preferential detection of histidine-tagged (His-tagged) recombinant fusion proteins in protein polyacrylamide gels.

The unique fluorescent stain specifically detects histidine-tagged fusion proteins that have been electrophoresed in polyacrylamide gels (e.g., SDS-PAGE). With this stain, expression of His-tagged recombinant proteins can be assessed directly in-gel, eliminating membrane transfer and Western blotting steps that are typically used to confirm purity and yield of tagged proteins. Although the lower limit of detection depends on which UV lamp (300nm) and detection equipment (CCD camera best) are used, the kit is convenient for in-process assessment of His-tagged protein production and is compatible with subsequent staining with coomassie dye and other total protein stains.

Highlights:

  • Two- to three-times faster than Western blotting – get results sooner and save hours of valuable research time

  • Detects directly on the gel – can eliminate the need for a membrane transfer and Western blot step for the detection of at least 0.2μg of 6xHis-tagged protein

  • Ready-to-use, two-reagent formula – no mixing, no diluting and no fuss; guarantees a simple-to-perform, mistake-free detection protocol

  • Specific fluorescent detection of 6xHis-tagged proteins – see only what you want to see (CCD camera recommended for low abundance proteins; UV-transilluminator for abundant proteins)

  • Compatible with double-staining – follow His-tagged protein staining with Thermo Scientific GelCode Blue Stain Reagent for a total protein profile determination

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store components at room temperature. Product is shipped at ambient temperature
Description Pierce 6xHis Protein Tag Stain Reagent Set
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye Proprietary Mix
Quantity 1 L
Target Molecule Proteins (His-Tagged)
Product Line Pierce
Product Type His Tag Protein Stain Set

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.