missing translation for 'onlineSavingsMsg'
Learn More

Pierce-1 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18433707 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433707 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433707 Supplier Novus Biologicals Supplier No. NBP281981IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Pierce-1 Polyclonal antibody specifically detects Pierce-1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pierce-1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias C9orf116, chromosome 9 open reading frame 116, FLJ13945, hypothetical protein LOC138162, MGC29761, Pierce 1, RbEST47, RP11-426A6.4
Host Species Rabbit
Immunogen Pierce-1 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human Pierce-1. The immunogen is located within the first 50 amino acids of Pierce-1. Amino Acid Squence: APKATAPPERTSDYYR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 138162
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.