missing translation for 'onlineSavingsMsg'
Learn More

PIDD Antibody (AL233) - BSA Free, Novus Biologicals™

Product Code. 18395253 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395253 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395253 Supplier Novus Biologicals Supplier No. NBP311679

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LRDD Polyclonal specifically detects LRDD in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, N/A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRDD
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Clone AL233
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin, N/A
Formulation PBS containing 10% glycerol
Gene Alias leucine-rich repeat and death domain-containing protein, leucine-rich repeats and death domain containing, MGC16925, p53-induced protein with a death domain, PIDD
Host Species Rabbit
Immunogen Recombinant human LRDD (Death Domain: aa 776-910).
Purification Method Epitope affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55367
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.