missing translation for 'onlineSavingsMsg'
Learn More

Pictilisib dimethanesulfonate, MedChemExpress

Product Code. 16453937
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16453937

Brand: Medchem Express HY20180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pictilisib dimethanesulfonate (GDC-0941 dimethanesulfonate) is a potent inhibitor of PI3Kα/δ with IC50 of 3 nM, with modest selectivity against p110β (11-fold) and p110γ (25-fold).

Pictilisib dimethanesulfonate (GDC-0941 dimethanesulfonate) is a potent inhibitor of PI3Kα/δ with IC50 of 3 nM, with modest selectivity against p110β (11-fold) and p110γ (25-fold).

Specifications

Chemical Name or Material Pictilisib dimethanesulfonate
CAS 957054-33-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H35N7O9S4
Quantity 50 mg
Synonym GDC-0941 dimethanesulfonateGDC-0941 2 MeSO3H salt
Solubility Information DMSO : 50 mg/mL (70.84 mM; Need ultrasonic)
SMILES O=S(N1CCN(CC1)CC2=CC3=C(S2)C(N4CCOCC4)=NC(C5=CC=CC6=C5C=NN6)=N3)(C)=O.O=S(O)(C)=O.O=S(O)(C)=O
Molecular Weight (g/mol) 705.85
Formula Weight 705.85
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.