missing translation for 'onlineSavingsMsg'
Learn More

Picolinic Acid, TRC

Product Code. 17980405
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17980405 1 g 1g
17901795 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17980405 Supplier Toronto Research Chemicals Supplier No. TRCP4372201G
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-98-6
Molecular Formula C6 H5 N O2
Molecular Weight (g/mol) 123.11
Synonym Picolinic acid (7CI,8CI), 2-Carboxypyridine, 2-Pyridylcarboxylic acid, NSC 171, PCL 016, Pyridine-2-carboxylic acid, o-Pyridinecarboxylic acid, α-Pyridinecarboxylic acid, Picolonic Acid, Picolinic acid (7CI,8CI), 2-Carboxypyridine, 2-Pyridylcarboxylic acid, NSC 171, PCL 016, Pyridine-2-carboxylic acid, o-Pyridinecarboxylic acid, α-Pyridinecarboxylic acid, Picolonic Acid
IUPAC Name pyridine-2-carboxylic acid
SMILES OC(=O)c1ccccn1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H5NO2/c8-6(9)5-3-1-2-4-7-5/h1-4H,(H,8,9)
Formula Weight 123.032
Percent Purity >95%
Chemical Name or Material Picolinic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.