missing translation for 'onlineSavingsMsg'
Learn More

Picolinamide, TRC

Product Code. 30415672
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30415672 10 g 10g
30409842 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30415672 Supplier Toronto Research Chemicals Supplier No. TRCP43700010G
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1452-77-3
Molecular Formula C6 H6 N2 O
Molecular Weight (g/mol) 122.12
Synonym 2-Pyridinecarboxamide, Picolinamide (6CI,7CI,8CI), 2-Carbamoylpyridine, NSC 524473, Picolinic acid amide, Picolinoylamide, Pyridine-2-carboxamide, α-(Aminocarbonyl)pyridine, α-Picolinamide, α-Picolinic acid amide, α-Pyridinecarboxamide
IUPAC Name pyridine-2-carboxamide
SMILES NC(=O)c1ccccn1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H6N2O/c7-6(9)5-3-1-2-4-8-5/h1-4H,(H2,7,9)
Formula Weight 122.048
Chemical Name or Material Picolinamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.