missing translation for 'onlineSavingsMsg'
Learn More

Picolinamide, 98%

Product Code. 15438330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15438330 10 g 10g
15740879 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438330 Supplier Thermo Scientific Acros Supplier No. 443360100
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1452-77-3 | C6H6N2O | 122.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1452-77-3
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD00023483
InChI Key IBBMAWULFFBRKK-UHFFFAOYSA-N
Synonym picolinamide, 2-pyridinecarboxamide, picolinoylamide, 2-carbamoylpyridine, picolinic acid amide, 2-picolinamide, 2-aminocarbonyl-pyridine, alpha-picolinamide, pyridinecarboxamide, unii-i3550ccl59
PubChem CID 15070
ChEBI CHEBI:8200
IUPAC Name pyridine-2-carboxamide
SMILES NC(=O)C1=CC=CC=N1

Specifications

Melting Point 106.0°C to 109.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 10 g
Formula Weight 122.13
Percent Purity 98%
Chemical Name or Material Picolinamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.