missing translation for 'onlineSavingsMsg'
Learn More

Picloram, 95%

Product Code. 15478159
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15478159 25 g 25g
15468159 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478159 Supplier Thermo Scientific Alfa Aesar Supplier No. J66702.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA) | CAS: 1918-02-1 | C6H3Cl3N2O2 | 241.45 g/mol

Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-1-1918
Molecular Formula C6H3Cl3N2O2
Molecular Weight (g/mol) 241.45
MDL Number MFCD00012101
InChI Key NQQVFXUMIDALNH-UHFFFAOYSA-N
Synonym picloram, 4-amino-3,5,6-trichloropicolinic acid, tordon, borolin, grazon, amdon, 2-pyridinecarboxylic acid, 4-amino-3,5,6-trichloro, k-pin, tordon 10k, tordon 22k
PubChem CID 15965
ChEBI CHEBI:34922
IUPAC Name 4-amino-3,5,6-trichloropyridine-2-carboxylic acid
SMILES C1(=C(C(=NC(=C1Cl)Cl)C(=O)O)Cl)N

Specifications

Melting Point 219°C
Boiling Point 421°C
Flash Point >110°C (230°F)
Packaging Plastic bottle
Quantity 25 g
Beilstein 479075
Solubility Information Soluble in acetone
Formula Weight 241.46
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Picloram
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.