missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Picloram 100 ug/mL in Methanol, Dr. Ehrenstorfer
Spécification
Spécification
| Chemical Name or Material | Picloram |
| CAS | 2-1-1918 |
| Analyte or Component Names | Picloram |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H3 Cl3 N2 O2 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | 2-Pyridinecarboxylic acid, 4-amino-3,5,6-trichloro-, Picolinic acid, 4-amino-3,5,6-trichloro- (8CI), 4-Amino-3,5,6-trichloro-2-pyridinecarboxylic acid, 3,5,6-Trichloro-4-aminopicolinic acid, 4-Amino-3,5,6-trichloro-2-picolinic acid, 4-Amino-3,5,6-trichloropicolinic acid, 4-Amino-3,5,6-trichloropyridine-2-carboxylic acid, 4-Aminotrichloropicolinic acid, ATCP, Grazon PC, NSC 233899, Padron, Padron (herbicide), Picloram, Tordon |
| InChI Formula | InChI=1S/C6H3Cl3N2O2/c7-1-3(10)2(8)5(9)11-4(1)6(12)13/h(H2,10,11)(H,12,13) |
| SMILES | Nc1c(Cl)c(Cl)nc(C(=O)O)c1Cl |
| Afficher plus |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?