missing translation for 'onlineSavingsMsg'
Learn More

Piclamilast, Tocris Bioscience™

Product Code. 13464884
Encompass
Click to view available options
10mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective PDE4 inhibitor

Potent and selective inhibitor of phosphodiesterase (PDE) 4 (IC50 = 1 nM at PDE4 from human neutrophils). Displays >19,000-fold selectivity over other PDE isoenzymes. Exhibits anti-inflammatory effects in models of allergic inflammation. Also inhibits LTB4 synthesis in human neutrophils (IC50 = 2 nM).

Specifications

CAS 144035-83-6
Molecular Formula C18H18Cl2N2O3
Quantity 10mg
Synonym piclamilast, cpodpmb, 3-cyclopentyloxy-n-3,5-dichloropyridin-4-yl-4-methoxybenzamide, unii-wm58d7c3zt, 3-cyclopentyloxy-n-3,5-dichloro-4-pyridinyl-4-methoxybenzamide, wm58d7c3zt, benzamide, 3-cyclopentyloxy-n-3,5-dichloro-4-pyridinyl-4-methoxy, chembl42126, 3-cyclopentyloxy-n-3,5-dichloro-4-pyridyl-p-anisamide, 3-cyclopentyloxy-n-3,5-dichloro-4-pyridyl-4-methoxybenzamide
InChI Key RRRUXBQSQLKHEL-UHFFFAOYSA-N
SMILES COC1=C(C=C(C=C1)C(=O)NC2=C(C=NC=C2Cl)Cl)OC3CCCC3
IUPAC Name 3-cyclopentyloxy-N-(3,5-dichloropyridin-4-yl)-4-methoxybenzamide
Molecular Weight (g/mol) 381.253
PubChem CID 154575
ChEBI CHEBI:47619
Formula Weight 381.25
Percent Purity >99%
Chemical Name or Material Piclamilast
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'