Learn More
PICK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1073
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-073 detects protein interacting with protein kinase C (PICK1) from mouse cells. PA1-073 has been successfully used in Western blot, immunocytochemistry, and immunoprecipitation procedures. By Western blot, this antibody detects an ∽52 kDa protein representing PICK1 from AtT20 cell extract. Immunocytochemical staining of CV1 cells transfected with PICK1 results in a diffuse staining pattern when expressed alone and coclustered when coexpressed with mGluR7. The PA1-073 immunogen is a synthetic peptide corresponding to residues M(1) F A D L D Y D I E E D K L G I P T(18) of mouse PICK1. The PA1-073 immunizing peptide (Cat. # PEP-133) is available for use in neutralization and control experiments.
PSD-95/DLG/ZO-1 (PDZ) domain-containing proteins play a central role in synaptic membrane protein localization. The protein interacting with protein kinase C (PICK1) is a synaptic PDZ domain protein that also contains a coiled-coil and acidic domain. Studies indicate that PICK1 functions as a targeting and transport protein and has been shown to interact with protein kinase C alpha (PKCalpha), AMPA-type glutamate receptors, and several other membrane receptors via its PDZ domain. The interaction of PICK1 with PKCalpha is highly dependent on the activation of the kinase. It appears that PICK1 directs the activated form of PKCalpha to membrane anchored GluR2. Once phosphorylated by PKCalpha, GluR2 is released from the synaptic anchor proteins. Once released, GluR2 is transported from the synaptic membrane in a PICK1-dependent manner.Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
PICK1 | |
Polyclonal | |
Unconjugated | |
Pick1 | |
MGC15204; OTTHUMP00000028509; PICK; Pick1; PRKCA-binding protein; Prkcabp; Protein interacting with C kinase 1; protein interacting with PRKCA 1; protein kinase C, alpha binding protein; protein kinase C-alpha-binding protein; protein that interacts with C kinase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18693, 84591 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Q62083, Q9EP80 | |
Pick1 | |
Synthetic peptide corresponding to residues M(1) F A D L D Y D I E E D K L G I P T(18) of mouse PICK1. | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |