missing translation for 'onlineSavingsMsg'
Learn More

Piccolo Antibody [DyLight 755], Novus Biologicals™

Product Code. 30733368 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733368 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733368 Supplier Novus Biologicals Supplier No. AF7935Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Piccolo Polyclonal antibody specifically detects Piccolo in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Piccolo
Applications Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias piccolo (presynaptic cytomatrix protein), protein piccolo
Host Species Sheep
Immunogen E. coli-derived recombinant human Piccolo, Ala4065-Phe4328, Accession # NP_149015
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 27445
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.