missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PICALM Polyclonal specifically detects PICALM in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Dilution | Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of human CALM. The exact sequence is proprietary. |
| Molecular Weight of Antigen | 71 kDa |
| Purification Method | Affinity Purified |
| Regulatory Status | RUO |
| Isotype | IgG |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?