missing translation for 'onlineSavingsMsg'
Learn More

PIBF1 Antibody (7A3), Novus Biologicals™

Product Code. 18340389 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340389 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340389 Supplier Novus Biologicals Supplier No. H00010464M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PIBF1 Monoclonal antibody specifically detects PIBF1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIBF1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 7A3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_006337
Gene Alias C13orf24, chromosome 13 open reading frame 24, KIAA1008, PIBF, progesterone immunomodulatory binding factor 1, progesterone-induced blocking factor 1, progesterone-induced-blocking factor 1, RP11-505F3.1
Host Species Mouse
Immunogen PIBF1 (NP_006337, 660 a.a. ~ 755 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EKSALLQTKNQMALDLEQLLNHREELAAMKQILVKMHSKHSENSLLLTKTEPKHVTENQKSKTLNVPKEHEDNIFTPKPTLFTKKEAPEWSKKQKM
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10464
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.