missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIAS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585183
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (81%), Chicken (93%). Positive Control: Target recombinant protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein involved in the regulation of transcription factors involved in MAP kinase signaling. The symbol MIZ1 has also been associated with ZBTB17 which is a different gene located on chromosome 1. Two alternatively spliced transcripts encoding different isoforms have been described.
Specifications
| PIAS2 | |
| Polyclonal | |
| Unconjugated | |
| Pias2 | |
| 6330408K17Rik; AI462206; androgen receptor-interacting protein 3; ARIP3; AU018068; DAB2-interacting protein; Dib; DIP; E3 SUMO-protein ligase PIAS2; E3 SUMO-protein transferase PIAS2; MGC102682; MIZ; Miz1; Msx-interacting zinc finger protein; Msx-interacting-zinc finger; msx-interacting-zinc finger protein; Pias2; PIAS-NY protein; Piasx; PIASxalpha; PIASX-ALPHA; PIASxalpha6; PIASxb; PIASxbeta; PIASX-BETA; protein inhibitor of activated STAT 2; protein inhibitor of activated STAT x; protein inhibitor of activated STAT, 2; Protein inhibitor of activated STAT2; RING-type E3 ubiquitin transferase PIAS2; SIZ2; zinc finger, MIZ-type containing 4; ZMIZ4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9063 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.01 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O75928 | |
| Pias2 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human PIAS2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction