missing translation for 'onlineSavingsMsg'
Learn More

PIAS2 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18418264 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418264 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418264 Supplier Novus Biologicals Supplier No. NBP177159AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIAS2 Polyclonal antibody specifically detects PIAS2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIAS2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias Androgen receptor-interacting protein 3, ARIP3, DAB2-interacting protein, DIP, E3 SUMO-protein ligase PIAS2, MGC102682, miz, Miz1, Msx-interacting zinc finger protein, Msx-interacting-zinc finger, PIAS-NY protein, PIASX, PIASX-ALPHA, PIASX-BETA, Protein inhibitor of activated STAT x, protein inhibitor of activated STAT, 2, Protein inhibitor of activated STAT2, SIZ2, zinc finger, MIZ-type containing 4, ZMIZ4
Host Species Rabbit
Immunogen Antibody was raised against a 18 amino acid synthetic peptide near the amino terminus of human PIAS2. The immunogen is located within amino acids 110 - 160 of PIAS2. Amino Acid Squence: DTKPTFEMQQPSPPIPPV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 9063
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.