missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIAS1 Recombinant Rabbit Monoclonal Antibody (JE36-84)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544844
This item is not returnable.
View return policy
Description
PIAS1 Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow
PIAS1 functions as an E3-type small ubiquitin-like modifier (SUMO) ligase, stabilizing the interaction between UBE2I and the substrate, and as a SUMO-tethering factor. This protein plays a crucial role in transcriptional coregulation of various cellular pathways, including the STAT pathway, the p53 pathway and the steroid hormone signaling pathway. It functions in testis as a nuclear receptor transcriptional coregulator and may have a role in androgen receptor initiation and maintenance of spermatogenesis. The effects of transcriptional coregulation, transactivation or silencing, may vary depending upon the biological context.
Specifications
| PIAS1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O75925, O88907 | |
| PIAS1 | |
| Synthetic peptide within human PIAS1 aa 71-120/651. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| JE36-84 | |
| Unconjugated | |
| PIAS1 | |
| 2900068C24Rik; AR interacting protein; Ddxbp1; DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1; DEAD/H box-binding protein 1; E3 SUMO-protein ligase PIAS1; E3 SUMO-protein transferase PIAS1; GBP; Gu binding protein; GU/RH-II; gu-binding protein; PIAS1; protein inhibitor of activated STAT 1; protein inhibitor of activated STAT protein 1; protein inhibitor of activated STAT, 1; protein inhibitor of activated STAT-1; RING-type E3 ubiquitin transferase PIAS1; RNA helicase II-binding protein; zinc finger, MIZ-type containing 3; ZMIZ3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 56469, 8554 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction