missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIAS1/PIAS2 Recombinant Rabbit Monoclonal Antibody (24GB920)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | PIAS1/PIAS2 |
|---|---|
| Clone | 24GB920 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275679
![]() |
Invitrogen™
MA557171 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276376
|
Invitrogen™
MA557172 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PIAS1/PIAS2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a member of the protein inhibitor of activated STAT (PIAS) family. PIAS proteins function as SUMO E3 ligases and play important roles in many cellular processes by mediating the sumoylation of target proteins. This protein plays a central role as a transcriptional coregulator of numerous cellular pathways includign the STAT1 and nuclear factor kappaB pathways. Alternate splicing results in multiple transcript variants. [provided by RefSeq, Mar 2016]Specifications
| PIAS1/PIAS2 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| PIAS1 | |
| 2900068C24Rik; 6330408K17Rik; AI462206; androgen receptor-interacting protein 3; AR interacting protein; ARIP3; AU018068; DAB2-interacting protein; Ddxbp1; DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1; DEAD/H box-binding protein 1; Dib; DIP; E3 SUMO-protein ligase PIAS1; E3 SUMO-protein ligase PIAS2; E3 SUMO-protein transferase PIAS1; E3 SUMO-protein transferase PIAS2; GBP; GU/RH-II; Gu-binding protein; MIZ; Miz1; msx-interacting zinc finger protein; Msx-interacting-zinc finger; msx-interacting-zinc finger protein; PIAS1; Pias2; PIAS-NY protein; PIASX; PIASxalpha; PIASX-ALPHA; PIASxalpha6; PIASxb; PIASxbeta; PIASX-BETA; protein inhibitor of activated STAT 1; protein inhibitor of activated STAT 2; protein inhibitor of activated STAT protein 1; Protein inhibitor of activated STAT x; protein inhibitor of activated STAT, 1; protein inhibitor of activated STAT, 2; protein inhibitor of activated STAT-1; Protein inhibitor of activated STAT2; RING-type E3 ubiquitin transferase PIAS1; RING-type E3 ubiquitin transferase PIAS2; RNA helicase II-binding protein; SIZ2; zinc finger, MIZ-type containing 3; zinc finger, MIZ-type containing 4; ZMIZ3; ZMIZ4 | |
| PIAS1, PIAS2 | |
| Primary | |
| Affinity chromatography |
| 24GB920 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75925, O75928, O88907, Q6AZ28, Q8C5D8 | |
| 17344, 300772, 56469, 83422, 8554, 9063 | |
| A synthesized peptide derived from human PIAS1 (aa 600-651). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title