missing translation for 'onlineSavingsMsg'
Learn More

PIAS1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18406672 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406672 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406672 Supplier Novus Biologicals Supplier No. NBP177158AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIAS1 Polyclonal antibody specifically detects PIAS1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIAS1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias AR interacting protein, DDXBP1MGC141878, DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1, DEAD/H box-binding protein 1, E3 SUMO-protein ligase PIAS1, GBPRNA helicase II-binding protein, GU/RH-II, Gu-binding protein, MGC141879, Protein inhibitor of activated STAT protein 1, protein inhibitor of activated STAT, 1, protein inhibitor of activated STAT-1, zinc finger, MIZ-type containing 3, ZMIZ3
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human PIAS1. The immunogen is located within the last 50 amino acids of PIAS1. Amino Acid Squence: SNSLRESHSHTVTNR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8554
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.