missing translation for 'onlineSavingsMsg'
Learn More

PIANO Paraffins Standard

Product Code. 30327382
Change view
Click to view available options
Quantity:
1 x 1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327382 1 x 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327382 Supplier VHG Labs Supplier No. VHGPIANOPAR1
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Nonane∣n-Octane∣n-Pentadecane∣n-Pentane∣n-Tetradecane∣n-Tridecane∣n-Undecane
CAS 124-18-5, 112-40-3, 142-82-5, 110-54-3, 111-84-2, 111-65-9
Analyte or Component Names n-Decane∣n-Dodecane∣n-Heptane∣n-Hexane∣n-Nonane∣n-Octane∣n-Pentadecane∣n-Pentane∣n-Tetradecane∣n-Tridecane∣n-Undecane
Shipping Condition Room Temperature
Solution Type Mixture
Solvent or Matrix Hydrocarbons
Industry Type Petrochemical Testing
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) n-Pentadecane 7.0999 wt%∣n-Tetradecane 8.8164 wt%∣n-Tridecane 8.9365 wt%∣n-Nonane 9.0525 wt%∣n-Dodecane 9.193 wt%∣n-Decane 9.2594 wt%∣n-Undecane 9.3183 wt%∣n-Pentane 9.4016 wt%∣n-Hexane 9.5293 wt%∣n-Octane 9.5602 wt%∣n-Heptane 9.833 wt%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.