missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIANO Isoparaffins Mixture 90, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 2,2,3-Trimethylbutane∣2,2,3-Trimethylpentane∣2,2-Dimethylhexane∣2,2-Dimethyloctane∣2,2-Dimethylpentane∣2,3-Dimethylbutane∣2,3-Dimethylheptane∣2,3-Dimethylhexane∣2,3-Dimethyloctane∣2,3-Dimethylpentane∣2,4-Dimethylhexane∣2,4-Dimethylpentane∣2,5-Dimethylheptane∣2,5-Dimethylhexane∣2-Methylbutane∣2-Methylheptane∣2-Methylhexane∣2-Methylnonane∣2-Methyloctane∣2-Methylpentane∣3,3-Diethylpentane∣3,3-Dimethylheptane∣3,3-Dimethyloctane∣3,3-Dimethylpentane∣3,4-Dimethylheptane∣3,5-Dimethylheptane∣3-Ethylhexane∣3-Ethyloctane∣3-Ethylpentane∣3-Methylheptane∣3-Methylhexane∣3-Methylnonane∣3-Methyloctane∣3-Methylpentane∣4-Methylheptane |
| CAS | 464-06-2, 564-02-3, 590-73-8, 15869-87-1, 590-35-2, 79-29-8 |
| Analyte or Component Names | 2,2,3-Trimethylbutane∣2,2,3-Trimethylpentane∣2,2-Dimethylhexane∣2,2-Dimethyloctane∣2,2-Dimethylpentane∣2,3-Dimethylbutane∣2,3-Dimethylheptane∣2,3-Dimethylhexane∣2,3-Dimethyloctane∣2,3-Dimethylpentane∣2,4-Dimethylhexane∣2,4-Dimethylpentane∣2,5-Dimethylheptane∣2,5-Dimethylhexane∣2-Methylbutane∣2-Methylheptane∣2-Methylhexane∣2-Methylnonane∣2-Methyloctane∣2-Methylpentane∣3,3-Diethylpentane∣3,3-Dimethylheptane∣3,3-Dimethyloctane∣3,3-Dimethylpentane∣3,4-Dimethylheptane∣3,5-Dimethylheptane∣3-Ethylhexane∣3-Ethyloctane∣3-Ethylpentane∣3-Methylheptane∣3-Methylhexane∣3-Methylnonane∣3-Methyloctane∣3-Methylpentane∣4-Methylheptane |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 2,3-Dimethylbutane 0.4 %∣3-Ethylpentane 0.5 %∣3-Ethylhexane 0.7 %∣3,5-Dimethylheptane 0.8 %∣3,3-Dimethylpentane 1.2 %∣2,2-Dimethylhexane 1.3 %∣2,3-Dimethylheptane 1.5 %∣2,3-Dimethylhexane 1.6 %∣2,4-Dimethylhexane 1.6 %∣3,3-Diethylpentane 1.6 %∣2,2,3-Trimethylpentane 1.7 %∣3,3-Dimethylheptane 1.7 %∣3-Methylhexane 1.7 %∣2,2-Dimethylpentane 1.8 %∣2,3-Dimethylpentane 1.8 %∣2-Methylbutane 2.2 %∣2-Methylhexane 2.2 %∣4-Methylheptane 3.2 %∣2-Methylpentane 3.3 %∣3,3-Dimethyloctane 3.3 %∣2,2-Dimethyloctane 3.4 %∣2,4-Dimethylpentane 3.7 %∣2,5-Dimethylhexane 3.7 %∣2-Methylnonane 3.7 %∣3,4-Dimethylheptane 3.7 %∣3-Ethyloctane 3.7 %∣2-Methyloctane 3.8 %∣2,2,3-Trimethylbutane 3.9 %∣2,3-Dimethyloctane 3.9 %∣2-Methylheptane 4.4 %∣3-Methylpentane 5.4 %∣3-Methylheptane 5.5 %∣3-Methyloctane 5.6 %∣2,5-Dimethylheptane 5.7 %∣3-Methylnonane 5.8 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction