missing translation for 'onlineSavingsMsg'
Learn More

PIANO Aromatics Standard

Product Code. 30322645
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30322645 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322645 Supplier VHG Labs Supplier No. VHGPIANOARO0.1
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,4,5-Tetramethylbenzene∣1,2,4-Triethylbenzene∣1,2,4-Trimethylbenzene∣1,2-Diethylbenzene∣1,2-Dimethyl-3-ethylbenzene∣1,2-Dimethyl-4-ethylbenzene∣1,3,5-Triethylbenzene∣1,3-Dimethyl-5-ethylbenzene∣1,4-Dimethyl-2-ethylbenzene∣1-Methyl-2-ethylbenzene∣1-Methyl-2-isopropylbenzene∣1-Methyl-2-n-propylbenzene∣1-Methyl-3-isopropylbenzene∣1-Methyl-3-n-propylbenzene∣1-Methyl-4-n-propylbenzene∣2-Ethyl-m-xylene∣2-Methylbutylbenzene∣2-tert-Butyltoluene∣3-Ethyltoluene∣4-Cymene∣4-Ethyltoluene∣5-tert-Butyl-m-Xylene∣Benzene∣Ethylbenzene∣Isobutylbenzene∣Isopropylbenzene∣m-Xylene (1,3-Dimethylbenzene)∣n-Butylbenzene∣n-Hexylbenzene∣n-Pentylbenzene∣o-Xylene (1,2-Dimethylbenzene)∣p-(tert-Butyl)ethylbenzene∣Propylbenzene∣p-Xylene (1,4-Dimethylbenzene)∣sec-Butylbenzene∣tert-Butylbenzene∣Toluene
CAS 95-93-2, 877-44-1, 95-63-6, 135-01-3, 933-98-2, 934-80-5
Analyte or Component Names 1,2,4,5-Tetramethylbenzene∣1,2,4-Triethylbenzene∣1,2,4-Trimethylbenzene∣1,2-Diethylbenzene∣1,2-Dimethyl-3-ethylbenzene∣1,2-Dimethyl-4-ethylbenzene∣1,3,5-Triethylbenzene∣1,3-Dimethyl-5-ethylbenzene∣1,4-Dimethyl-2-ethylbenzene∣1-Methyl-2-ethylbenzene∣1-Methyl-2-isopropylbenzene∣1-Methyl-2-n-propylbenzene∣1-Methyl-3-isopropylbenzene∣1-Methyl-3-n-propylbenzene∣1-Methyl-4-n-propylbenzene∣2-Ethyl-m-xylene∣2-Methylbutylbenzene∣2-tert-Butyltoluene∣3-Ethyltoluene∣4-Cymene∣4-Ethyltoluene∣5-tert-Butyl-m-Xylene∣Benzene∣Ethylbenzene∣Isobutylbenzene∣Isopropylbenzene∣m-Xylene (1,3-Dimethylbenzene)∣n-Butylbenzene∣n-Hexylbenzene∣n-Pentylbenzene∣o-Xylene (1,2-Dimethylbenzene)∣p-(tert-Butyl)ethylbenzene∣Propylbenzene∣p-Xylene (1,4-Dimethylbenzene)∣sec-Butylbenzene∣tert-Butylbenzene∣Toluene
Shipping Condition Room Temperature
Solution Type Mixture
Solvent or Matrix Hydrocarbons
Industry Type Petrochemical Testing
Quantity 0.1 mL
Concentration or Composition (by Analyte or Components) 1,2,4,5-Tetramethylbenzene 0.2334 wt%∣2-tert-Butyltoluene 0.7514 wt%∣1,2,4-Triethylbenzene 1.0448 wt%∣4-Cymene 1.0535 wt%∣1,2-Diethylbenzene 1.0781 wt%∣1-Methyl-3-isopropylbenzene 1.0904 wt%∣1-Methyl-2-isopropylbenzene 1.1078 wt%∣2-Methylbutylbenzene 1.1238 wt%∣2-Ethyl-m-xylene 1.1286 wt%∣1-Methyl-3-n-propylbenzene 2.0728 wt%∣5-tert-Butyl-m-Xylene 2.1242 wt%∣1,2-Dimethyl-3-ethylbenzene 2.1488 wt%∣3-Ethyltoluene 2.1537 %∣n-Butylbenzene 2.1689 wt%∣Isopropylbenzene 2.184 wt%∣p-(tert-Butyl)ethylbenzene 2.1906 wt%∣1-Methyl-4-n-propylbenzene 2.1938 wt%∣sec-Butylbenzene 2.1999 wt%∣o-Xylene (1,2-Dimethylbenzene) 2.2103 wt%∣1,2-Dimethyl-4-ethylbenzene 2.2152 wt%∣1,3-Dimethyl-5-ethylbenzene 2.2236 wt%∣1-Methyl-2-n-propylbenzene 2.2239 wt%∣1,4-Dimethyl-2-ethylbenzene 2.2242 wt%∣m-Xylene (1,3-Dimethylbenzene) 2.2242 wt%∣4-Ethyltoluene 2.3067 wt%∣1,2,4-Trimethylbenzene 2.4894 wt%∣1-Methyl-2-ethylbenzene 2.5376 wt%∣Isobutylbenzene 4.3442 wt%∣n-Pentylbenzene 4.397 wt%∣Propylbenzene 4.4182 wt%∣n-Hexylbenzene 4.4219 wt%∣tert-Butylbenzene 4.4477 wt%∣1,3,5-Triethylbenzene 4.4837 wt%∣Toluene 4.489 wt%∣p-Xylene (1,4-Dimethylbenzene) 4.7269 wt%∣Ethylbenzene 6.64 wt%∣Benzene 7.0606 wt%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.