missing translation for 'onlineSavingsMsg'
Learn More

PI4KIIIbeta-IN-9, MedChemExpress

Product Code. p-200098415
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497908

Brand: Medchem Express HY19798

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PI4KIIIbeta-IN-9 is a potent PI4KIIIβ inhibitor with an IC50 of 7 nM. PI4KIIIbeta-IN-9 also inhibits PI3Kδ and PI3Kγ with IC50s of 152 nM and 1046 nM, respectively.

PI4KIIIbeta-IN-9 is a potent PI4KIIIβ inhibitor with an IC50 of 7 nM. PI4KIIIbeta-IN-9 also inhibits PI3Kδ and PI3Kγ with IC50s of 152 nM and 1046 nM, respectively.

Specifications

Chemical Name or Material PI4KIIIbeta-IN-9
CAS 1429624-84-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H25N3O5S2
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (205.09 mM; Need ultrasonic)
SMILES CC(N=C1NC(C2CCCC2)=O)=C(S1)C3=CC=C(C(S(=O)(NC4=CC=C(O)C=C4)=O)=C3)OC
Molecular Weight (g/mol) 487.59
Formula Weight 487.59
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.