missing translation for 'onlineSavingsMsg'
Learn More

PI4KIII beta inhibitor 3, MedChemExpress

Product Code. 16440256
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
Unit Size:
10mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16440256

Brand: Medchem Express HY15679

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PI4KIII beta inhibitor 3 is a novel and high effective PI4KIIIβ inhibitor with IC50 of 5.7 nM.

PI4KIII beta inhibitor 3 is a novel and high effective PI4KIIIβ inhibitor with IC50 of 5.7 nM.

Specifications

Chemical Name or Material PI4KIII beta inhibitor 3
Physical Form Solid
CAS 1245319-54-3
Color Light Yellow
Quantity 2 mg
Purity Grade Notes Research
Molecular Formula C22H22N8OS
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 20 mg/mL (44.79 mM; Need ultrasonic)
SMILES O=C(N1CCN(C2=C3C(SC(C4=CC=CN=C4)=N3)=NC(N)=N2)CC1)NC5=CC=C(C)C=C5
Molecular Weight (g/mol) 446.53
Formula Weight 446.53
Percent Purity 99.44%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.