missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PI4KB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544560
This item is not returnable.
View return policy
Description
Peptide sequence: LILSDELKPA HRKRELPSLS PAPDTGLSPS KRTHQRSKSD ATASISLSSN Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
PI4KB and PI4KA are the two human type III PI4 kinases, which share sequence similarly with PI3 kinases and phosphorylate the 4 position of phosphotidyl inositol. Transporter for alanine, serine, cysteine, and threonine. Exhibits sodium dependence. Target protein phosphorylates phosphatidylinositol (PtdIns) at the D4 position of the inositol ring; may play a role in neuronal differentiation and maturation.
Specifications
| PI4KB | |
| Polyclonal | |
| Unconjugated | |
| Pi4kb | |
| catalytic phosphatidylinositol 4-kinase beta; ESTM41; lipid kinase; DrPI4K III beta; NPIK; phosphatidylinositol 4-kinase; phosphatidylinositol 4-kinase beta; phosphatidylinositol 4-kinase III beta; phosphatidylinositol 4-kinase, catalytic, beta; phosphatidylinositol 4-kinase, catalytic, beta polypeptide; phosphatidylinositol 4-kinase, wortmannin-sensitive; PI4K92; PI4KB; PI4Kbeta; PI4K-beta; PI4KIII; PI4KIIIBETA; PI4KIII-beta; Pik4cb; PtdIns 4-kinase beta; type III phosphatidylinositol 4-kinase beta; zgc:152715 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5298 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UBF8 | |
| Pi4kb | |
| Synthetic peptide directed towards the N-terminal of human PI4KB (aa 239-288). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction