missing translation for 'onlineSavingsMsg'
Learn More

PI4KB/PI4KIII beta Antibody, DyLight 405, Novus Biologicals™

Product Code. 18430874 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430874 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430874 Supplier Novus Biologicals Supplier No. NBP212814V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PI4KB/PI4KIII beta Polyclonal antibody specifically detects PI4KB/PI4KIII beta in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI4KB/PI4KIII beta
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias DKFZp686O1820, EC 2.7.1, EC 2.7.1.67, FLJ30129, phosphatidylinositol 4-kinase, catalytic, beta, pi4K92, PI4Kbeta, PI4K-beta, PI4KIIIBETA, type III phosphatidylinositol 4-kinase beta, wortmannin-sensitive
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Phosphatidylinositol 4-Kinase, Catalytic, beta using the numbering given in entry CAA09495.1 (GeneID 5298).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5298
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.