missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PI4KB (PI4K beta), GST Tag Recombinant Protein

Product Code. 15478783
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
Each
This item is not returnable. View return policy

Product Code. 15478783

Brand: Invitrogen™ PV5278

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

PI4KB and PI4KA are the two human type III PI4 kinases, which share sequence similarly with PI3 kinases and phosphorylate the 4 position of phosphotidyl inositol. Transporter for alanine, serine, cysteine, and threonine. Exhibits sodium dependence. Target protein phosphorylates phosphatidylinositol (PtdIns) at the D4 position of the inositol ring; may play a role in neuronal differentiation and maturation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UBF8
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 5298
Molecular Weight (g/mol) 120 kDa
Name Human PI4KB (PI4K beta), GST Tag
Quantity 1 mg
Regulatory Status RUO - research use only
Gene Alias catalytic phosphatidylinositol 4-kinase beta; ESTM41; lipid kinase; DrPI4K III beta; NPIK; phosphatidylinositol 4-kinase; phosphatidylinositol 4-kinase beta; phosphatidylinositol 4-kinase III beta; phosphatidylinositol 4-kinase, catalytic, beta; phosphatidylinositol 4-kinase, catalytic, beta polypeptide; phosphatidylinositol 4-kinase, wortmannin-sensitive; PI4K92; PI4KB; PI4Kbeta; PI4K-beta; PI4KIII; PI4KIIIBETA; PI4KIII-beta; Pik4cb; PtdIns 4-kinase beta; type III phosphatidylinositol 4-kinase beta; zgc:152715
Common Name PI4KB
KinaseFamily PI3 Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.