Learn More
PI4KA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12411-1-AP-20UL
 View more versions of this product
Product Code. 16810124
  • £135.00 / 20µL
Description
This gene encodes a phosphatidylinositol (PI) 4-kinase which catalyzes the first committed step in the biosynthesis of phosphatidylinositol 4,5-bisphosphate. The mammalian PI 4-kinases have been classified into two types, II and III, based on their molecular mass, and modulation by detergent and adenosine. The protein encoded by this gene is a type III enzyme that is not inhibited by adenosine. Two transcript variants encoding different isoforms have been described for this gene.
Specifications
Specifications
PI4KA
Polyclonal
Unconjugated
PI4KA
PI4 kinase alpha, PI4K ALPHA, pi4K230, PI4KA, PIK4, PIK4CA, PtdIns 4 kinase alpha
Rabbit
Antigen Affinity Chromatography
RUO
224020, 5297, 64161
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
E9Q3L2, O08662, P42356
PI4KA
PI4KA Fusion Protein Ag3062
20 μL
Primary
Rat, Mouse, Human