missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PI4KA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109770
This item is not returnable.
View return policy
Description
Immunogen sequence: PEAIVLDIDY KSGTPMQSAA KAPYLAKFKV KRCGVSELEK EGLRCRSDSE DECSTQEADG QKISWQAAIF KVGDDCRQDM L.
PI4KB and PI4KA are the two human type III PI4 kinases, which share sequence similarly with PI3 kinases and phosphorylate the 4 position of phosphotidyl inositol. The mammalian PI 4-kinases have been classified into two types, II and III, based on their molecular mass, and modulation by detergent and adenosine. The protein encoded by PI4KA is a type III enzyme that is not inhibited by adenosine. Two transcript variants encoding different isoforms have been described for PI4KA.
Specifications
| PI4KA | |
| Polyclonal | |
| Unconjugated | |
| PI4KA | |
| leuserpin 2; phosphatidylinositol 4-kinase; phosphatidylinositol 4-kinase 230; phosphatidylinositol 4-kinase a; phosphatidylinositol 4-kinase alpha; phosphatidylinositol 4-kinase, catalytic, alpha; phosphatidylinositol 4-kinase, catalytic, alpha polypeptide; phosphatidylinositol 4-kinase, type III, alpha; pi4K230; Pi4ka; PI4K-ALPHA; PI4-kinase alpha; Pik4; PIK4CA; PMGYCHA; ptdIns-4-kinase alpha; testicular secretory protein Li 35 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 224020, 5297, 64161 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.60 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| E9Q3L2, O08662, P42356 | |
| PI4KA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1780-1860 of human PI4KA (P42356). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction