missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI4K2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15074-1-AP-20UL
This item is not returnable.
View return policy
Description
Phosphatidylinositol 4-kinases (PI4Ks) phosphorylate phosphatidylinositol to generate phosphatidylinositol 4-phosphate (PIP), an immediate precursor of several important signaling and scaffolding molecules. PIP itself may also have direct functional and structural roles. PI4K2B is a primarily cytosolic PI4K that is recruited to membranes, where it stimulates phosphatidylinositol 4, 5-bisphosphate synthesis (Wei et al., 2002 ).Specifications
| PI4K2B | |
| Polyclonal | |
| Unconjugated | |
| PI4K2B | |
| PI 4 Kinase type 2 beta, PI4K2B, PI4KII BETA, PI4KIIB, PIK42B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 305419, 55300, 67073 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5XIL2, Q8CBQ5, Q8TCG2 | |
| PI4K2B | |
| PI4K2B Fusion Protein Ag7120 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction