missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PI4K2A (PI4K2 alpha), GST Tag Recombinant Protein

Product Code. 17168755
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
20µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17168755 100 μg 100µg
15950313 20 μg 20µg
17138805 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17168755 Supplier Invitrogen™ Supplier No. A32905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This PI4K2A (PI4K2 alpha) recombinant human protein (full length) was expressed in insect cells. PI4K2A (PI4K2 alpha) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for PI4K2A (PI4K2 alpha) is NP_060895.1.

PI4K2A (PI4K2 alpha) lipid kinase or phosphatidylinositol 4-kinase type 2 alpha is involved in the formation of phosphatidylinositolpolyphosphates (PtdInsPs) which are centrally involved in many biologic processes ranging from cell growth and organization of the actin cytoskeleton to endo- and exocytosis. PI4K2A phosphorylates PtdIns at the D-4 position which is an essential step in the biosynthesis of PtdInsPs. PI4K2A is a novel regulator of tumor growth by its action on angiogenesis and HIF-1alpha regulation. PI4K2A generates PtdIns4P-rich domains within the Golgi which regulate targeting of clathrin adaptor AP-1 complexes. PI4K2A is also required for Wnt to regulate LRP6 phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BTU6
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.1mM EDTA, 0.1mM PMSF, 150mM NaCl, 10mM glutathione, 0.25mM DTT, 25% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 55361
Molecular Weight (g/mol) 80.3 kDa
Name Human PI4K2A (PI4K2 alpha), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 55 kDa type II phosphatidylinositol 4-kinase; null; phosphatidylinositol 4-kinase; phosphatidylinositol 4-kinase type 2 alpha; phosphatidylinositol 4-kinase type 2-alpha; phosphatidylinositol 4-kinase type II; phosphatidylinositol 4-kinase type II (PI4KII); phosphatidylinositol 4-kinase type II-alpha; Pi4k2; PI4K2A; Pi4kii; PIK42A
Common Name PI4K2A
Gene Symbol Pi4k2a
KinaseFamily PI3 Kinase
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.