Learn More
Invitrogen™ Human PI4K2A (PI4K2 alpha), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This PI4K2A (PI4K2 alpha) recombinant human protein (full length) was expressed in insect cells. PI4K2A (PI4K2 alpha) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for PI4K2A (PI4K2 alpha) is NP_060895.1.
Specifications
Specifications
| Accession Number | Q9BTU6 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris HCl with 0.1mM EDTA, 0.1mM PMSF, 150mM NaCl, 10mM glutathione, 0.25mM DTT, 25% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 55361 |
| Molecular Weight (g/mol) | 80.3 kDa |
| Name | Human PI4K2A (PI4K2 alpha), GST Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 20 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.