missing translation for 'onlineSavingsMsg'
Learn More

PI4K2A Antibody (3E1), Novus Biologicals™

Product Code. 18383839 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383839 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383839 Supplier Novus Biologicals Supplier No. H00055361M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PI4K2A Monoclonal antibody specifically detects PI4K2A in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI4K2A
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3E1
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_060895
Gene Alias DKFZp761G1923, EC 2.7.1.67, phosphatidylinositol 4-kinase type 2 alpha, phosphatidylinositol 4-kinase type 2-alpha, phosphatidylinositol 4-kinase type II (PI4KII), Phosphatidylinositol 4-kinase type II-alpha, PI4KII, PIK42A, RP11-548K23.6
Host Species Mouse
Immunogen PI4KII (NP_060895.1, 383 a.a. ~ 477 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LILPKISDPNFVKDLEEDLYELFKKDPGFDRGQFHKQIAVMRGQILNLTQALKDNKSPLHLVQMPPVIVETARSHQRSSSESYTQSFQSRKPFFS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55361
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.