missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PI3K p55 gamma Recombinant Rabbit Monoclonal Antibody (K01_2W51)

Product Code. 30827044
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827044 100 μL
30825859 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827044 Supplier Invitrogen™ Supplier No. MA564295

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

PI3K p55 gamma Recombinant Monoclonal Antibody for Western Blot

PIK3R3 binds to activated (phosphorylated) protein-tyrosine kinases through its SH2 domain and regulates their kinase activity. During insulin stimulation, it also binds to IRS-1. It is a component of a heterodimer of p110 (catalytic) and p55 (regulatory) subunits. The protein is expressed at highest levels in brain and testis. Lower levels are detected in adipose tissue, kidney, heart, lung and skeletal muscle. The protein contains 2 SH2 domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI3K p55 gamma
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2W51
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Pik3r3
Gene Accession No. Q63789, Q64143, Q92569
Gene Alias AA414954; p55; p55-GAMMA; p55pik; phosphatidylinositol 3 kinase, regulatory subunit, polypeptide 3 (p55); phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma; phosphatidylinositol 3-kinase p55 subunit; phosphatidylinositol 3-kinase regulatory subunit gamma; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 3 (p55, gamma); phosphoinositide-3-kinase regulatory subunit 3; phosphoinositide-3-kinase, regulatory subunit 3 (gamma); PI3K regulatory subunit gamma; PI3-kinase p85 subunit gamma; PI3-kinase regulatory subunit gamma; PI3-kinase subunit p55-gamma; Pik3r3; ptdIns-3-kinase p85-gamma; ptdIns-3-kinase regulatory subunit gamma; ptdIns-3-kinase regulatory subunit p55-gamma; RP4-533D7.2
Gene Symbols Pik3r3
Host Species Rabbit
Immunogen A synthesized peptide derived from human PI 3 Kinase p55 gamma.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18710, 60664, 8503
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.