missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI3K p110(gamma) Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20662-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that belongs to the pi3/pi4-kinase family of proteins. The gene product is an enzyme that phosphorylates phosphoinositides on the 3-hydroxyl group of the inositol ring. It is an important modulator of extracellular signals, including those elicited by E-cadherin-mediated cell-cell adhesion, which plays an important role in maintece of the structural and functional integrity of epithelia. In addition to its role in promoting assembly of adherens junctions, the protein is thought to play a pivotal role in the regulation of cytotoxicity in NK cells. The gene is located in a commonly deleted segment of chromosome 7 previously identified in myeloid leukemias.Specifications
| PI3K p110(gamma) | |
| Polyclonal | |
| Unconjugated | |
| PIK3CG | |
| p120 PI3K, PI3 kinase subunit gamma, PI3CG, PI3K, PI3K gamma, PI3K p110(gamma), PI3Kgamma, PIK3, PIK3CG, PtdIns 3 kinase subunit gamma | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5294 | |
| -20°C | |
| Liquid |
| Immunoprecipitation | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P48736 | |
| PIK3CG | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction