missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI3K p110(beta) Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20584-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to PIK3CB.
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.Specifications
| PI3K p110(beta) | |
| Polyclonal | |
| Unconjugated | |
| PIK3CB | |
| p110 BETA, PI3 Kinase p110 beta, PI3 kinase subunit beta, PI3K, PI3K beta, PI3K p110(beta), PI3Kbeta, PI3KCB, PIK3C1, PIK3CB, PtdIns 3 kinase subunit beta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5291 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P42338 | |
| PIK3CB | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction