missing translation for 'onlineSavingsMsg'
Learn More

PI3K-IN-1, MedChemExpress

Product Code. 16484968
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484968 10 mg 10mg
16451217 50 mg 50mg
16447707 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16484968 Supplier Medchem Express Supplier No. HY12068

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PI3K-IN-1 (XL-147 derivative 1) is a potent inhibitor of PI3K. PI3K-IN-1 (25 μM) blocks PI3K/Akt signaling pathways.

PI3K-IN-1 (XL-147 derivative 1) is a potent inhibitor of PI3K. PI3K-IN-1 (25 μM) blocks PI3K/Akt signaling pathways.

Specifications

Chemical Name or Material PI3K-IN-1
CAS 1349796-36-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H29N5O6S
Quantity 10 mg
Synonym XL-147 derivative 1
Solubility Information DMSO : 27.5 mg/mL (45.86 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES COC1=CC(NC2=NC3=CC=CC=C3N=C2NS(C4=CC=C(C=C4)NC(C5=CC=C(C(OC)=C5)C)=O)(=O)=O)=CC(OC)=C1
Molecular Weight (g/mol) 599.66
Formula Weight 599.66
Percent Purity 96.34%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.