missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX PI3 Kinase (p110α/p85α) Human Protein

Product Code. 17300414
Encompass_Preferred
Click to view available options
Quantity:
20 μg
250 μg
Unit Size:
20µg
250µg
This item is not returnable. View return policy

Product Code. 17300414

Brand: Eurofins DiscoverX 14602

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active complex of N-terminal His6-tagged recombinant full-length human p110α and untagged, recombinant, full length, human p85α, for use in Kinase Assays.

Phosphatidylinositol 3-kinase is composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit. The protein encoded by this gene represents the catalytic subunit, which uses ATP to phosphorylate PtdIns, PtdIns4P and PtdIns(4,5)P2. This gene has been found to be oncogenic and has been implicated in cervical cancers.

GenBank U79143 / GenBank XM_043865

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 125.3 kDa (P110α) and 83.7 kDa (p85α)
Quantity 20 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His / Untagged
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Full-length
Purity or Quality Grade Combined ≥66%
Protein PI3 Kinase (p110α/p85α), active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.