Learn More
Eurofins DiscoverX PI3 Kinase (p110α (H1047R)/p85α) Mouse Protein
Description
The H1047R substitution in p110α is associated with a number of human cancers. The residue is conserved between the human and mouse sequences. In vitro studies have shown that this mutation confers increased lipid kinase activity compared to wild type, and is able to induce oncogenic transformation.
GenBank BC089038 / GenBank NM_001077495
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 129 kDa (p110α(H1047R)) and 83.6 kDa (p85α) |
| Quantity | 10 μg |
| Species | Murine |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His / Untagged |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | Combined ≥65% |
| Protein | PI 3-Kinase (p110α(H1047R)/p85α), murine |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.