missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PI3 Kinase p110 Alpha Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27921-1-AP-20UL
This item is not returnable.
View return policy
Description
The PI3 Kinase p110 Alpha antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PI3 Kinase p110 Alpha. This antibody recognizes human, mouse, rat antigen. The PI3 Kinase p110 Alpha antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| PI3 Kinase p110 Alpha | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5290 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, ELISA | |
| 700 μg/mL | |
| MGC142161; MGC142163; PI3K; p110-alpha | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 110 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction