missing translation for 'onlineSavingsMsg'
Learn More

PI3 Kinase p110 Alpha Mouse anti-Human, Unconjugated, Clone: 1F6A7, Proteintech

Mouse Monoclonal Antibody

£109.00 - £274.38

Specifications

Antigen PI3 Kinase p110 Alpha
Clone 1F6A7
Concentration 1000 μg/mL
Applications Co-immunoprecipitation (Co-IP), Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16829485
View Documents
Proteintech
67071-1-IG-20UL
20 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16819485
View Documents
Proteintech
67071-1-IG-150UL
150 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

The PI3 Kinase p110 Alpha antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PI3 Kinase p110 Alpha. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The PI3 Kinase p110 Alpha antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Specifications

PI3 Kinase p110 Alpha
1000 μg/mL
Monoclonal
Mouse
5290
Primary
1F6A7
Co-immunoprecipitation (Co-IP), Immunohistochemistry
Unconjugated
Human, Mouse, Rat, Pig, Rabbit
IgG2b
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.