missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PI16 Recombinant Rabbit Monoclonal Antibody (9Z5V7)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | PI16 |
|---|---|
| Clone | 9Z5V7 |
| Concentration | 0.072 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30283460
|
Invitrogen™
MA558211 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30284194
![]() |
Invitrogen™
MA558212 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: LTEDEKQTMV DLHNQYRAQV SPPASDMLQM RWDDELAAFA KAYAQKCVWG HNKERGRRGE NLFAITDEGM DVPLAVGNWH EEHEYYNFST ATCDPNQMCG HYTQVVWSKT ERIGCGSHFC ETLQGVEEAN IHLLVCNYEP PGNVKGRKPY QEGTPCSQCP LGYSCENSLC EPMRNPEKAQ D.
May inhibit cardiomyocyte growth. [UniProt]Specifications
| PI16 | |
| 0.072 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q9ET66 | |
| 74116 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-210 of mouse CD364/PI16 (NP_076223.3). | |
| Antibody |
| 9Z5V7 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| PI16 | |
| 1200009H11Rik; CD364; Cripi; CRISP9; CRISP-9; Cysteine-rich protease inhibitor; cysteine-rich secretory protein 9; microseminoprotein, beta-binding protein; MSMBBP; peptidase inhibitor 16; Pi16; PI-16; protease inhibitor 16; PSEC0164; PSP94-binding protein; PSPBP; UNQ289/PRO328 | |
| PI16 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title