missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PI16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531881
This item is not returnable.
View return policy
Description
Recommended positive controls: A549. Predicted reactivity: Rhesus Monkey (97%), Chimpanzee (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Peptidase inhibitor 16 (PI16), also known as CRISP-9 or PSPBP, is a member of the cysteine-rich secretory protein (CRISP) family and plays a significant role in various biological processes. This protein consists of 436 amino acids and features three N-linked glycosylation motifs, suggesting its localization to the plasma membrane through a GPI anchor. PI16 has been implicated as a binding protein for prostate secretory protein of 94 amino acids (PSP94), which reflects its potential involvement in reproductive biology. Recent findings highlight PI16's expression in fibroblasts, particularly marking a precursor fibroblast cell state, which is critical in fibroblast lineage development. This expression is heightened in certain pathological conditions, such as neuropathic pain and inflammatory arthritis. In neuropathic pain models such as the spared nerve injury (SNI) model, elevated PI16 levels are noted in fibroblasts within the dorsal root ganglia (DRG) and sciatic nerve, but not in neurons or glia. Interestingly, mice lacking PI16 are protected against neuropathic pain, indicating its role in enhancing pain signaling. Conversely, in rheumatoid arthritis (RA), PI16's overexpression in peripheral blood and synovial fluid correlates with worsened arthritis severity, partly by suppressing the expression of the regulatory T cell marker Foxp3. This complex role of PI16, affecting various signaling pathways and cellular states, underscores its potential as a therapeutic target in pain and inflammatory diseases.
Specifications
| PI16 | |
| Polyclonal | |
| Unconjugated | |
| PI16 | |
| 1200009H11Rik; CD364; Cripi; CRISP9; CRISP-9; Cysteine-rich protease inhibitor; cysteine-rich secretory protein 9; microseminoprotein, beta-binding protein; MSMBBP; peptidase inhibitor 16; Pi16; PI-16; protease inhibitor 16; PSEC0164; PSP94-binding protein; PSPBP; UNQ289/PRO328 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 221476 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q6UXB8 | |
| PI16 | |
| Recombinant protein encompassing a sequence within the center region of human PI16. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction