Learn More
Invitrogen™ PI15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114174
Description
Immunogen sequence: PPTNNFTDIE AALKAQLDSA DIPKARRKRY ISQNDMIAIL DYHNQVRGKV For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 91%.
This gene encodes a trypsin inhibitor. The protein shares similarity to insect venom allergens, mammalian testis-specific proteins and plant pathogenesis-related proteins. It is frequently expressed in human neuroblastoma and glioblastoma cell lines, and thus may play a role in the central nervous system. [provided by RefSeq].
Specifications
| PI15 | |
| Polyclonal | |
| Unconjugated | |
| PI15 | |
| 25 kDa trypsin inhibitor; CRISP8; CRISP-8; Cysteine-rich secretory protein 8; DKFZp686F0366; P24TI; P25TI; Peptidase inhibitor 15; PI15; PI-15; protease inhibitor 15; sugarCrisp | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51050 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O43692 | |
| PI15 | |
| Synthetic peptide directed towards the N terminal region of human PI15 (aa 32-81). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.