missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PI15 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA552312

Product Code. 15770261

  • £472.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LLFSLLCEAS TVVLLNSTDS SPPTNNFTDI EAALKAQLDS ADIPKARRKR YISQNDMIAI LDYHNQVRGK VFPPAANMEY MVWDENLAKS AEAWAATCIW DHGPSYLLRF LGQNLSVRTG RYRSILQLVK PWYDEVKDY Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.

This gene encodes a trypsin inhibitor. The protein shares similarity to insect venom allergens, mammalian testis-specific proteins and plant pathogenesis-related proteins. It is frequently expressed in human neuroblastoma and glioblastoma cell lines, and thus may play a role in the central nervous system. [provided by RefSeq].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PI15
Polyclonal
Unconjugated
PI15
25 kDa trypsin inhibitor; CRISP8; CRISP-8; Cysteine-rich secretory protein 8; DKFZp686F0366; P24TI; P25TI; Peptidase inhibitor 15; PI15; PI-15; protease inhibitor 15; sugarCrisp
Rabbit
Antigen Affinity Chromatography
RUO
51050
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O43692
PI15
Recombinant protein corresponding to Human PI15. Recombinant protein control fragment (Product #RP-89267).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.