missing translation for 'onlineSavingsMsg'
Learn More

Cytiva pI Markers

Product Code. 10024594 Shop All Cytiva Products
Change view
Click to view available options
pH Range:
2.5 to 6.5
3 to 10
5 to 10.5
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
10024594 2.5 to 6.5 Each
10616515 5 to 10.5 Each
10649084 3 to 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10024594 Supplier Cytiva Supplier No. 17047201

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Run these markers in parallel with the experimental samples on isoelectric focusing gels. Cytiva Life Sciences™ pI Markers are lyophilized mixtures of stable, salt-free, highly purified proteins.

The markers may be run in parallel with the experimental samples on isoelectric focusing gels and are not prestained.

  • The precisely defined pI of the markers allows a permanent and accurate calibration of sample components
  • pH gradients and pI can be measured to within 0.01 to 0.05 pH unit
  • The progress of the focusing experiment can be monitored visually by the focusing of methyl red or Cytochrome c

Not for use in denaturing (urea) systems.

TRUSTED_SUSTAINABILITY

Specifications

Color White
pH Range 2.5 to 6.5
Form Lyophilized
For Use With (Application) To be used for isoelectric focusing (IEF)
Kit Contents Pepsinogen 2.80 pI (Native), Amylglucosidase 3.50 pI (Native), Methyl red (dye) 3.75 pI (Native), Glucose Oxidase 4.25 pI (Native), Trypsin inhibitor 4.55 pI (Native), bLactoglobulin A 5.20 pI (Native), Carbonic Anhydrase B (Bovine) 5.85 pI (Native)
Quantity 1 Kit: 10 Vials
Storage Requirements 4°C
Product Type pl Marker Kit
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.