missing translation for 'onlineSavingsMsg'
Learn More

PI 3-Kinase p85 beta Antibody (PIK3R2/292), DyLight 680, Novus Biologicals™

Product Code. 18364126 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364126 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364126 Supplier Novus Biologicals Supplier No. NBP314138FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PI 3-Kinase p85 beta Monoclonal antibody specifically detects PI 3-Kinase p85 beta in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI 3-Kinase p85 beta
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PIK3R2/292
Conjugate DyLight 680
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias P85B, phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta, phosphatidylinositol 3-kinase regulatory subunit beta, phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta), phosphoinositide-3-kinase, regulatory subunit 2 (beta), phosphoinositide-3-kinase, regulatory subunit 2 (p85 beta), phosphoinositide-3-kinase, regulatory subunit, polypeptide 2 (p85 beta), PI3K regulatory subunit beta, PI3-kinase regulatory subunit beta, PI3-kinase subunit p85-beta, ptdIns-3-kinase regulatory subunit beta, ptdIns-3-kinase regulatory subunit p85-beta
Host Species Mouse
Immunogen Recombinant fragment of human PI 3-Kinase p85 beta protein (around aa 520-720) (exact sequence is proprietary) (Uniprot: O00459)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5296
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.