missing translation for 'onlineSavingsMsg'
Learn More

PI 3-Kinase p85 alpha Antibody (572106) [DyLight 650], Novus Biologicals™

Product Code. 30735437 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735437 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735437 Supplier Novus Biologicals Supplier No. FAB2998W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PI 3-Kinase p85 alpha Monoclonal antibody specifically detects PI 3-Kinase p85 alpha in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI 3-Kinase p85 alpha
Applications Western Blot
Classification Monoclonal
Clone 572106
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias AGM7, GRB1, IMD36, p85, p85-ALPHA, Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha, phosphatidylinositol 3-kinase regulatory subunit alpha, phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha), phosphatidylinositol 3-kinase-associated p-85 alpha, phosphoinositide-3-kinase regulatory subunit alpha, phosphoinositide-3-kinase, regulatory subunit 1 (alpha), PI3K regulatory subunit alpha, PI3-kinase regulatory subunit alpha, PI3-kinase subunit p85-alpha, PIK3R1, PtdIns-3-kinase regulatory subunit alpha, PtdIns-3-kinase regulatory subunit p85-alpha
Host Species Mouse
Immunogen E. coli-derived recombinant human PI 3-Kinase p85 alpha , aa Lys519-Gly644, Accession # P27986
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Immune Dysfunction, Lipid and Metabolism, mTOR Pathway, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5295
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.